missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAI16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £299.00
Specifications
| Antigen | RAI16 |
|---|---|
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18305896
|
Novus Biologicals
NBP3-15603-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18398196
|
Novus Biologicals
NBP3-15603-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAI16 Polyclonal antibody specifically detects RAI16 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
| RAI16 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| family with sequence similarity 160, member B2, FLJ11125, FLJ21801, MGC138352, RAI16, retinoic acid induced 16, Retinoic acid-induced protein 16 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 74-303 of human RAI16 (NP_073586.5). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS, 50% glycerol, pH7.3 | |
| 64760 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title