Learn More
Invitrogen™ RAI16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116902
Description
Positive Samples: 293T, A-549, SH-SY5Y, Mouse testis, Rat testis Immunogen sequence: GEAGPCLEYL LQHKILETLC TLGKAEYPPG MRQQVFQFFS KVLAQVQHPL LHYLSVHRPV QKLLRLGGTA SGSVTEKEEV QFTTVLCSKI QQDPELLAYI LEGKKIVGRK KACGEPTALP KDTTSHGDKD CSHDGAPARP QLDGESCGAQ ALNSHMPAET EELDGGTTES NLITSLLGLC QSKKSRVALK AQENLLLLVS MASPAAATYL VQSSACCPAI VRHLCQLYRS.
FAM160B2, also named as RAI16, may play a role in early lens development in mammals and in upregulation of genes in CD34+ cells in coronary artery disease patients.
Specifications
| RAI16 | |
| Polyclonal | |
| Unconjugated | |
| FAM160B2 | |
| FAM160B2; family with sequence similarity 160 member B2; family with sequence similarity 160, member B2; FP13191; G430067P06Rik; protein FAM160B2; RAI16; retinoic acid induced 16; retinoic acid-induced protein 16 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 239170, 306015, 64760 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.86 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q80YR2, Q86V87 | |
| FAM160B2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 74-303 of human FAM160B2 (NP_0735865). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.