missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAGEF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-06551
This item is not returnable.
View return policy
Description
RAGEF2 Polyclonal specifically detects RAGEF2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| RAGEF2 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| DKFZp667N084, DKFZp686I15116, KIA001LB, PDZ domain-containing guanine nucleotide exchange factor 2, PDZ domain-containing guanine nucleotide exchange factor I, PDZ-GEF2RA-GEF-2PDZGEF2PDZ domain containing guanine nucleotide exchange factor (GEF) 2, RAGEF2, Rap guanine nucleotide exchange factor (GEF) 6, rap guanine nucleotide exchange factor 6 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 1:10-1:500 | |
| NP_057424.2 | |
| RAPGEF6 | |
| The immunogen recognized by this antibody maps to a region between residue 1551 and 1601 of human PDZ domain containing guanine nucleotide exchange factor (GEF) 2 using the numbering given in entry NP_057424.2 (GeneID 51735). | |
| 0.1 mL | |
| Signal Transduction | |
| 51735 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction