missing translation for 'onlineSavingsMsg'
Learn More

RAGEF2 Antibody (2C5), Novus Biologicals™

Product Code. 18328099 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328099 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328099 Supplier Novus Biologicals Supplier No. H00051735M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RAGEF2 Monoclonal antibody specifically detects RAGEF2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAGEF2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2C5
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/ Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_057424
Gene Alias DKFZp667N084, DKFZp686I15116, KIA001LB, PDZ domain-containing guanine nucleotide exchange factor 2, PDZ domain-containing guanine nucleotide exchange factor I, PDZ-GEF2RA-GEF-2PDZGEF2PDZ domain containing guanine nucleotide exchange factor (GEF) 2, RAGEF2, Rap guanine nucleotide exchange factor (GEF) 6, rap guanine nucleotide exchange factor 6
Host Species Mouse
Immunogen RAPGEF6 (NP_057424, 1012 a.a. ~ 1110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LFPVVKKDMTFLHEGNDSKVDGLVNFEKLRMISKEIRQVVRMTSANMDPAMMFRQRSLSQGSTNSNMLDVQGGAHKKRARRSSLLNAKKLYEDAQMARK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51735
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.