Learn More
RAGE Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1075
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-075 detects receptor for advanced glycation endproducts (RAGE) from mouse and rat samples. PA1-075 has been successfully used in Western blot and immunohistochemistry procedures. By Western blot, this antibody detects two bands in the 45 kDa range representing the RAGE protein pre and post-glycosylation in mouse lung extract. This antibody also detects an ∽25 kDa protein that is believed to be proteolytic degradation product. Immunohistochemical staining of RAGE in transgenic mouse retina results in staining of the retinal pigmented epithelium and photo receptor cell layers. The PA1-075 immunogen is a synthetic peptides corresponding to residues C(362) W R K R Q P R (R/L) E E R K A P E S Q E D(380) of rat RAGE. This sequence is 100% conserved in mouse and 70% conserved in humans. This peptide (Cat. # PEP-194) is available for use in neutralization and control experiments.
Able to phosphorylate several exogenous substrates and to undergo autophosphorylation.Specifications
RAGE | |
Polyclonal | |
Unconjugated | |
AGER | |
advanced glycation end product receptor; advanced glycation end-product receptor; advanced glycation end-products receptor; advanced glycosylation end product-specific receptor; advanced glycosylation end product-specific receptor variant 2; advanced glycosylation end product-specific receptor variant 3; advanced glycosylation end product-specific receptor variant 4; advanced glycosylation end product-specific receptor variant 5; advanced glycosylation end-product specific receptor; Ager; MAPK/MAK/MRK overlapping kinase; MOK; MOK protein kinase; RAGE; RAGE isoform NtRAGE-delta; RAGE isoform sRAGE-delta; RAGE/AGER; RAGE1; RAGE-1; RAGE-4 ORF3; receptor for advanced glycation endproducts; receptor for advanced glycation end-products variant 20; receptor for advanced glycosylation end products; receptor of advanced glycosylation end products of proteins; immunoglobulin superfamily; MHC class II; MHC class III; renal cell carcinoma antigen; renal tumor antigen 1; SCARJ1; sRAGE; STK30 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11596, 81722 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Q62151, Q63495 | |
AGER | |
Synthetic peptides corresponding to residues C(362) W R K R Q P R (R/L) E E R K A P E S Q E D(380) of rat RAGE. | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |