Learn More
RAG2 Mouse anti-Human, Unconjugated, Clone: 1B4E5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66998-1-IG-20UL
This item is not returnable.
View return policy
Description
The RAG2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAG2. This antibody recognizes human, pig antigen. The RAG2 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| RAG2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Pig |
| Western Blot, Immunofluorescence, ELISA | |
| 1B4E5 | |
| Unconjugated | |
| 20 μL | |
| 5897 | |
| IgG1 |