Learn More
RAG2 Mouse anti-Human, Unconjugated, Clone: 1B4E5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66998-1-IG-150UL
 View more versions of this product
Product Code. 16819355
  • £349.00 / 150µL
Description
The RAG2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAG2. This antibody recognizes human, pig antigen. The RAG2 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
RAG2
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Pig
Western Blot, Immunofluorescence, ELISA
1B4E5
Unconjugated
150 μL
5897
IgG1