missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAG2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £549.00
Specifications
| Antigen | RAG2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227156
|
Novus Biologicals
NBP3-38173-100ul |
100 μL |
£549.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227063
|
Novus Biologicals
NBP3-38173-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAG2 Polyclonal antibody specifically detects RAG2 in Human,Mouse samples. It is validated for ELISA,Western BlotSpecifications
| RAG2 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Immunology | |
| PBS (pH 7.3), 50% glycerol | |
| 5897 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| RAG-2, recombination activating gene 2, V(D)J recombination-activating protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 351-527 of human RAG2 (NP_000527.2).,, Sequence:, AEDDTNEEQTTFTNSQTSTEDPGDSTPFEDSEEFCFSAEANSFDGDDEFDTYNEDDEEDESETGYWITCCPTCDVDINTWVPFYSTELNKPAMIYCSHGDGHWVHAQCMDLAERTLIHLSAGSNKYYCNEHVEIARALHTPQRVLPLKKPPMKSLRKKGSGKILTPAKKSFLRRLFD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title