missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAG2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-36703-100ul
This item is not returnable.
View return policy
Description
RAG2 Polyclonal antibody specifically detects RAG2 in Mouse samples. It is validated for ELISA,Western Blot
Specifications
| RAG2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| RAG-2, recombination activating gene 2, V(D)J recombination-activating protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 258-527 of human RAG2 (NP_001230715.1).,, Sequence:, AEDDTNEEQTTFTNSQTSTEDPGDSTPFEDSEEFCFSAEANSFDGDDEFDTYNEDDEEDESETGYWITCCPTCDVDINTWVPFYSTELNKPAMIYCSHGDGHWVHAQCMDLAERTLIHLSAGSNKYYCNEHVEIARALHTPQRVLPLKKPPMKSLRKKGSGKILTPAKKSFLRRLFD | |
| 100 μL | |
| Adaptive Immunity, Immunology | |
| 5897 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction