missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAG1 Monoclonal Antibody (1D9)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA517163

Product Code. 15388110

  • £434.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA5-17163 targets RAG1 in indirect ELISA and WB applications and shows reactivity with Human samples. The MA5-17163 immunogen is purified recombinant fragment of human RAG1 (amino acids: 818-868) expressed in E. Coli. MA5-17163 detects RAG1 which has a predicted molecular weight of approximately 119kDa.

The protein encoded by this gene is involved in activation of immunoglobulin V-D-J recombination. The encoded protein is involved in recognition of the DNA substrate, but stable binding and cleavage activity also requires RAG2. Defects in this gene can be the cause of several diseases.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RAG1
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
P15918
Rag1
Purified recombinant fragment of human RAG1 (amino acids: 818-868) expressed in E. Coli.
100 μg
Primary
Human
Antibody
IgG1
ELISA, Western Blot
1D9
Unconjugated
Rag1
E3 ubiquitin-protein ligase RAG1; Endonuclease RAG1; MGC43321; Rag1; RAG-1; RAG-1 Antibody; recombinase activating gene 1; recombinase activating protein; recombination activating 1; recombination activating gene 1; recombination activating gene 1 Antibody; recombination activating gene-1; recombination activating protein 1; recombination activation protein 1; recombination-activating gene 1; RING finger protein 74; RING-type E3 ubiquitin transferase RAG1; RNF74; RNF74 Antibody; V(D)J recombination-activating protein 1
Mouse
Protein G
RUO
5896
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis