Learn More
Invitrogen™ Human RAF1 (cRAF) [Y340D, Y341D], GST Tag Recombinant Protein
Descripción
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Catalytic, Mutant.
Especificaciones
Especificaciones
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM tris with 50% glycerol, 2mM DTT, 0.5mM EDTA, 150mM NaCl, 0.02% Triton X-100 and no preservative; pH 7.5 |
| Molecular Weight (g/mol) | 66.4 kDa |
| Name | Human RAF1 (cRAF) [Y340D, Y341D], GST Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 10 μg |
| Storage Requirements | -80°C, Avoid Freeze/Thaw Cycles |
| Regulatory Status | RUO |
| Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.