missing translation for 'onlineSavingsMsg'
Learn More

Raf-1 Rabbit anti-Human, Clone: S05-9B1, Novus Biologicals™

Product Code. 18383685 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18383685 100 μg 100µL
18344715 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18383685 Supplier Novus Biologicals Supplier No. NBP314880100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Raf-1 Monoclonal antibody specifically detects Raf-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Raf-1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone S05-9B1
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 2.5 ug/ml, Immunofluorescence 20 ug/ml
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias cRaf, c-Raf, CRAFraf proto-oncogene serine/threonine protein kinase, EC 2.7.11, EC 2.7.11.1, NS5, Proto-oncogene c-RAF, RAF, RAF proto-oncogene serine/threonine-protein kinase, Raf-1Oncogene RAF1, v-raf-1 murine leukemia viral oncogene homolog 1
Host Species Rabbit
Immunogen A synthetic peptide of human Raf-1 (Uniprot#: P04049)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, mTOR Pathway, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5894
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.