missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Raf-1, p Ser43 Antibody (SR2182), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21574-100ul
This item is not returnable.
View return policy
Description
Raf-1 Monoclonal antibody specifically detects Raf-1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Raf-1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR2182 | |
| Western Blot 1:500-1:1000 | |
| cRaf, c-Raf, CRAFraf proto-oncogene serine/threonine protein kinase, EC 2.7.11, EC 2.7.11.1, NS5, Proto-oncogene c-RAF, RAF, RAF proto-oncogene serine/threonine-protein kinase, Raf-1Oncogene RAF1, v-raf-1 murine leukemia viral oncogene homolog 1 | |
| A synthesized peptide derived from human phospho-Raf-1 [Ser43] (Uniprot #: P04049) | |
| 100 μg | |
| Apoptosis, Cancer, mTOR Pathway, Neuroscience, Phospho Specific, Signal Transduction | |
| 5894 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction