missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Raf-1, p Ser301 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-305
This item is not returnable.
View return policy
Description
Raf-1 Polyclonal specifically detects Raf-1 in Human, Rat samples. It is validated for Western Blot.
Specifications
| Raf-1 (p Ser301) | |
| Polyclonal | |
| Western Blot 1:1000 | |
| P11345 | |
| RAF1 | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser301 conjugated to KLH. Accession # P11365 | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Specific for the ∼74k Raf-1 protein phosphorylated at Ser301. Immunolabeling of the Raf-1 protein band is blocked by lambda-phosphatase treatment | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| cRaf, c-Raf, CRAFraf proto-oncogene serine/threonine protein kinase, EC 2.7.11, EC 2.7.11.1, NS5, Proto-oncogene c-RAF, RAF, RAF proto-oncogene serine/threonine-protein kinase, Raf-1Oncogene RAF1, v-raf-1 murine leukemia viral oncogene homolog 1 | |
| Rabbit | |
| 74 kDa | |
| 0.1 mL | |
| Apoptosis, Cancer, mTOR Pathway, Neuroscience, Phospho Specific, Signal Transduction | |
| 5894 | |
| Human, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction