missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
RAE1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Tekniske data
| Antigen | RAE1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|
30229413
|
Novus Biologicals
NBP3-38315-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227242
|
Novus Biologicals
NBP3-38315-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
RAE1 Polyclonal antibody specifically detects RAE1 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotTekniske data
| RAE1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| dJ481F12.3, dJ800J21.1, FLJ30608, homolog of yeast Rae1 (Bharathi) mRNA-associated protein of 41 kDa (Kraemer), MGC117333, MGC126076, MGC126077, MIG14, migration-inducing gene 14, Mnrp41, mRNA export factor, mRNA export protein, mRNA-associated protein mrnp 41, mRNA-binding protein, 41-kD, MRNP41, RAE1 (RNA export 1, S.pombe) homolog, Rae1 protein homolog, RAE1 RNA export 1 homolog (S. pombe) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-368 of human RAE1 (NP_003601.1).,, Sequence:, MSLFGTTSGFGTSGTSMFGSATTDNHNPMKDIEVTSSPDDSIGCLSFSPPTLPGNFLIAGSWANDVRCWEVQDSGQTIPKAQQMHTGPVLDVCWSDDGSKVFTASCDKTAKMWDLSSNQAIQIAQHDAPVKTIHWIKAPNYSCVMTGSWDKTLKFWDTRSSNPMMVLQLPERCYCADVIYPMAVVATAERGLIVYQLENQPSEFRRIESPLKHQHRCVAIFKDKQNKPTGFALGSIEGRVAIHYINPPNPAKDNFTFKCHRSNGTNTSAPQDIYAVNGIAFHPVHGTLATVGSDGRFSFWDKDARTKLKTSEQLDQPISACCFNHNGNIFAYASSYDWSKGHEFYNPQKKNYIFLRNAAEELKPRNKK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 8480 | |
| IgG | |
| Affinity purified |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel