Learn More
Invitrogen™ RAE1 alpha/beta/gamma Monoclonal Antibody (199215)
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  Invitrogen™ MA561039
Product Code. 30533956
  • £490.00 / 500µg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
RAE1 (RNA Export 1) is a gene that encodes a protein involved in the nuclear export of mRNA in eukaryotic cells. The RAE1 protein is a key component of the mRNA export machinery, facilitating the transport of mRNA from the nucleus to the cytoplasm, where it can be translated into proteins. Variants of this gene, including RAE1 alpha, beta, and gamma, represent different isoforms or homologs that may have distinct roles or regulatory mechanisms in various tissues or developmental stages. Proper functioning of RAE1 is essential for maintaining normal cellular activities and gene expression. Disruptions or mutations in the RAE1 gene can lead to defects in mRNA export, potentially resulting in various diseases, including neurodevelopmental disorders and cancer. Research into RAE1 and its isoforms continues to be important for understanding the complexities of mRNA transport and its implications in health and disease.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RAE1 alpha/beta/gamma
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
BaF3 mouse pro-B cell line transfected with Rae-1 beta and Mouse myeloma cell line NS0-derived recombinant mouse Rae-1 delta.
500 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Flow Cytometry, Inhibition Assays
199215
Unconjugated
Rat
Protein A/G
RUO
Mouse
Antibody
IgG2a