Learn More
Thermo Scientific™ RadEye PRD4 and PRD-ER4 Personal Radiation Detector
Product Code. 18861074 Shop All Thermo Scientific Products
Change view
Click to view available options
Detector Type:
1 Low Dose Rate Detector, 1 High Dose Rate Detector
Description:
PRD-ER4 Charger/BTLE Package
Unit Size:
Each
Product Code. Detector Type Description Unit Size
18861074 1 Low Dose Rate Detector, 1 High Dose Rate Detector PRD-ER4 Charger/BTLE Package Each
1 options
Product Code. 18861074 Supplier Thermo Scientific™ Supplier No. 425069171
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Thermo Scientific™ RadEye™ PRD4 and PRD-ER4 Personal Radiation Detectors help users detect real radiation threats with high sensitivity while minimizing nuisance alarms that slow screening, response, and decision-making.
Thermo Scientific™ RadEye™ PRD4 and Thermo Scientific™ RadEye™ PRD-ER4 Personal Radiation Detectors help security, emergency response, and radiation safety teams screen people, vehicles, facilities, and materials with high sensitivity, fewer nuisance alarms, and greater confidence in the field. The RadEye PRD4 detector is used for basic screening and interdiction, whereas the RadEye PRD-ER4 detector extends PRD-style screening to higher dose-rate operation capability. Built with natural background rejection (NBR) technology, both the RadEye PRD4 and RadEye PRD-ER4 detectors help distinguish artificial radiation from naturally occurring radioactive material (NORM), thereby reducing time spent adjudicating alarms and improving screening throughput. For missions requiring an added layer of threat awareness, the RadEye PRD4 detector also provides neutron indication capability.
The RadyEye PRD and PRD-ER4 Personal Radiation Detectors enable accurate dose rate measurements across the broader gamma spectrum, providing a greater assurance of personal safety. The neutron indication capability of the RadEye PRD provides an additional layer of detection critical to determining the level of radiation threat.
  • Efficiency in the field
  • Larger display
  • Neutron indication capability
  • 5% improved sensitivity over previous model
  • Linear high dose rate detector
  • Easy information, smarter decisions
  • No retraining or relearning for infrequent users G
  • et results automatically, without the need to press buttons
  • Quickly guides you through next steps after an alarm
  • Simple 4 button design
  • Comprehensive data neatly organized and presented on screen
  • Rugged and practical
  • Can be worn in holster or standard service belt Small and lightweight
  • Long battery life
  • Drop resistant to 1.5m IP65
  • Can be operated in extreme temperatures
TRUSTED_SUSTAINABILITY
Specifications
Description PRD-ER4 Charger/BTLE Package
Product Type Personal Radiation Detector
Detector Type 1 Low Dose Rate Detector, 1 High Dose Rate Detector
Includes RadEye PRD-ER4, desktop charger, BTLE charging back, and batteries