missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RADIL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | RADIL |
|---|---|
| Concentration | 0.43 mg/mL |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16899804
|
Proteintech
20284-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16889804
|
Proteintech
20284-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Radil is a broadly expressed adapter protein that contains a RA-like domain, a PDZ domain and a DIL domain. Like RAPL and RIAM, Radil interacts preferentially with active GTP-bound Rap1, and overexpression of Radil enhances integrin-mediated adhesion. In addition, Radil knock down inhibits cell-substrate adhesion in HT1080, AD293, and NMuMG cells and results in migration defects in SW1353 and zebrafish neural crest cells. Radil regulates neutrophil adhesion by specifically enhancing beta2-integrin activation, as well as focal adhesion kinase (FAK) and paxillin phosphorylation.Specifications
| RADIL | |
| Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q96JH8 | |
| 55698 | |
| RADIL Fusion Protein Ag13996 | |
| Primary | |
| -20°C | |
| RADIL |
| 0.43 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| KIAA1849, RADIL | |
| RADIL | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title