missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RADIL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£378.00
Specifications
| Antigen | RADIL |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
RADIL Polyclonal specifically detects RADIL in Mouse samples. It is validated for Western Blot.Specifications
| RADIL | |
| Polyclonal | |
| Purified | |
| RUO | |
| NP_848817 | |
| 55698 | |
| Synthetic peptide directed towards the C terminal of mouse D930005D10RIK. Peptide sequence PEPSGLCGRAVLDGQRNGPGGLPGAVLEGDAIQDAEPPAEASSPSSSAED. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| FLJ10324, FLJ60975, KIAA1849MGC161589, Rap GTPase interactor, Ras association and DIL domains, ras-associating and dilute domain-containing protein, RASIP2 | |
| RADIL | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title