missing translation for 'onlineSavingsMsg'
Learn More

Radafaxine hydrochloride, MedChemExpress

Product Code. 16439887
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16439887

Brand: Medchem Express HY17590

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Radafaxine hydrochloride (GW-353162A) is a DAT (dopamine transporter) and NET(norepinephrine transporter) transporters inhibitor, and nAChR family modulator.

Radafaxine hydrochloride (GW-353162A) is a DAT (dopamine transporter) and NET(norepinephrine transporter) transporters inhibitor, and nAChR family modulator.

Specifications

Chemical Name or Material Radafaxine hydrochloride
Physical Form Solid
CAS 106083-71-0
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C13H19Cl2NO2
For Use With (Application) Neuroscience-Neuromodulation
Synonym GW-353162A BW-306U
Solubility Information DMSO : ≥ 38 mg/mL (130.05 mM) ∣H2O : 33.33 mg/mL (114.07 mM; Need ultrasonic)
SMILES O[C@]1(C2=CC=CC(Cl)=C2)[C@H](C)NC(C)(C)CO1.[H]Cl
Molecular Weight (g/mol) 292.2
Formula Weight 292.2
Percent Purity 99.88%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.