missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAD9B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100736
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RAD9B.
RAD9B gene ontology (GO) annotations include DNA repair; DNA replication; cellular response to ionizing radiation; intra-S DNA damage checkpoint; regulation of signal transduction by p53 class mediator.
Specifications
| RAD9B | |
| Polyclonal | |
| Unconjugated | |
| RAD9B | |
| A630082N15Rik; BC021784; Cell cycle checkpoint control protein RAD9B; cell cycle checkpoint control protein RAD9B homolog; DNA repair exonuclease rad9 homolog B; hRAD9B; mRAD9B; RAD9 checkpoint clamp component B; RAD9 homolog B; RAD9B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 144715, 231724 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6WBX7, Q6WBX8 | |
| RAD9B | |
| A synthesized peptide derived from human RAD9B(Accession Q6WBX8), corresponding to amino acid residues L370-A420. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction