missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD9B Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06241APC
This item is not returnable.
View return policy
Description
RAD9B Polyclonal antibody specifically detects RAD9B in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| RAD9B | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| cell cycle checkpoint control protein RAD9B, cell cycle checkpoint control protein RAD9B homolog, DNA repair exonuclease rad9 homolog B, FLJ40346, hRAD9B, MGC75426, RAD9 homolog B (S. pombe) | |
| Produced in rabbits immunized with E. coli-derived Human RAD9B fragment. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 144715 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction