missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD9A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13035-1-AP-20UL
This item is not returnable.
View return policy
Description
ZNF295 contains 1 BTB (POZ) domain and 8 C2H2-type zinc fingers. ZNF295 may be involved in transcriptional regulation.Specifications
| RAD9A | |
| Polyclonal | |
| Unconjugated | |
| RAD9A | |
| hRAD9, RAD9, RAD9 homolog A (S. pombe), RAD9A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5883 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99638 | |
| RAD9A | |
| RAD9A Fusion Protein Ag3687 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction