missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAD9 Monoclonal Antibody (6B5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA532968
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
RAD9 is a component of the 9-1-1 cell cycle checkpoint response complex that plays a major role in DNA repair. The 9-1-1 complex is recruited to DNA lesions upon damage by the RAD17-replication factor C clamp loader complex. RAD9 acts as a sliding clamp platform on DNA for several proteins involved in long-patch base excision repair.
Specifications
| RAD9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99638, Q9Z0F6 | |
| RAD9A | |
| Recombinant full length Human RAD9A. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| 6B5 | |
| Unconjugated | |
| RAD9A | |
| Cell cycle checkpoint control protein RAD9A; chromatin-binding protein RAD9; DNA repair exonuclease rad9 homolog A; DNA repair protein RAD9; hRAD9; mRAD9; QtsA-19913; Rad9; RAD9 checkpoint clamp component A; RAD9 homolog A; RAD9 homolog A (S. pombe); Rad9a; Rad9-like protein; rCG47324-like; unnamed protein product; YD9934.02C; YDR217C | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 100361529, 19367, 5883 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction