missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD54L2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | RAD54L2 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18619041
|
Novus Biologicals
NBP2-95145-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673242
|
Novus Biologicals
NBP2-95145-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAD54L2 Polyclonal antibody specifically detects RAD54L2 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| RAD54L2 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Chromatin Modifiers, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 23132 | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Androgen receptor-interacting protein 4, ARIP4, EC 3.6.1, EC 3.6.4.12, FLJ21396, FLJ22400, helicase ARIP4, KIAA0809HSPC325, RAD54-like 2 (S. cerevisiae), RAD54-like protein 2, SRISNF2L | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1178-1467 of human RAD54L2 (NP_055921.2). ELQQYADVAAARESRQSSPSTNAALPGPPAQLMDSSAVPGTALGTEPRLGGHCLNSSLLVTGQPCGDRHPVLDLRGHKRKLATPPAAQESSRRRSRKGHLPAPVQPYEHGYPVSGGFAMPPVSLNHNLTTPFTSQAGENSLFMGSTPSYYQLSNLLADARLVFPVTTDPLVPAGPVSSSSTATSVTASNPSFMLNPSVPGILPSYSLPFSQPLLSEPRMFAPFPSPVLPSNLSRGMSIYPGYMSPHAGYPAGGLLRSQVPPFDSHEVAEVGFSSNDDEDKDDDVIEVTGK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title