Learn More
RAD54B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541272
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: DAVLIVKGKS FILKNLEGKD IGRGIGYKFK ELEKIEEGQT LMICGKEIEV Sequence homology: Cow: 86%; Dog: 79%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 79%; Rat: 100%
RAD54B Polyclonal antibody specifically detects RAD54B in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
RAD54B | |
Polyclonal | |
Unconjugated | |
RAD54B | |
DNA repair and recombination protein RAD54B; E130016E03Rik; fibrinogen silencer-binding protein; FSBP; LOW QUALITY PROTEIN: DNA repair and recombination protein RAD54B; RAD54 homolog B; RAD54 homolog B (S. cerevisiae); RAD54, S. cerevisiae, homolog of, B; Rad54b; RDH54; RGD1306507 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
25788 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y620 | |
RAD54B | |
Synthetic peptide directed towards the N-terminal of human RAD54B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |