missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD54B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99021-50ul
This item is not returnable.
View return policy
Description
RAD54B Polyclonal antibody specifically detects RAD54B in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| RAD54B | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| DNA repair and recombination protein RAD54B, EC 3.6.1, EC 3.6.4.-, fibrinogen silencer binding protein, FSBP, RAD54 homolog B, RAD54 homolog B (S. cerevisiae), RDH54 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human RAD54B. | |
| 50 μg | |
| Cancer, DNA Repair, Homologous Recombination | |
| 25788 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction