missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAD52 Recombinant Rabbit Monoclonal Antibody (24GB5280)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557205
This item is not returnable.
View return policy
Description
RAD52 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
DNA double-strand breaks can be generated by exposure to ionizing radiation or through endogenous cellular reactions. Double-strand breaks are generally thought to be repaired via one of two distinct mechanisms: homologous recombination (HR) or non-homologous end joining (NHEJ). Homologous recombination requires the activity of the members of the RAD52 epistasis group, which includeRAD50, RAD51, RAD52, RAD54, RDH54/TID1, RAD55, RAD57, RAD59, MRE11, and XRS2. RAD52 can catalyze strand annealing reactions, and can form oligomers that attach to ends of single-stranded DNA.
Specifications
| RAD52 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| RAD52 | |
| DNA repair protein RAD52 homolog; RAD52; RAD52 homolog (S. cerevisiae); RAD52 homolog, DNA repair protein; Rad52yh; recombination protein RAD52; rhabdomyosarcoma antigen MU-RMS-40.23 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5893 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 24GB5280 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P43351 | |
| RAD52 | |
| A synthesized peptide derived from human RAD52 (aa 380-418). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction