missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD52 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | RAD52 |
|---|---|
| Clone | 3H8G9 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
Description
The RAD52 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAD52. This antibody recognizes human, mouse, rat antigen. The RAD52 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| RAD52 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 5893 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
| 3H8G9 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title