missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD52 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£292.00 - £451.00
Specifications
| Antigen | RAD52 |
|---|---|
| Dilution | Western Blot 0.04 - 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL, KnockDown Validated |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18219981
|
Novus Biologicals
NBP2-58116 |
100 μL |
£451.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18634717
|
Novus Biologicals
NBP2-58116-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAD52 Polyclonal specifically detects RAD52 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
| RAD52 | |
| Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown | |
| Unconjugated | |
| RUO | |
| Human | |
| 5893 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ILGGRDSHPAAGGGSVLCFGQCQYTAEEYQAIQKALRQRLGPEYISSRMAGGGQKVCYIEGHRVINLANEMFGYNGWAHSITQQNVDFVDLNNGKFYVGVCAFVRVQLKDGSYHEDVGYGVSE | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04 - 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL, KnockDown Validated | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, DNA Repair, Homologous Recombination | |
| DNA repair protein RAD52 homolog, RAD52 (S. cerevisiae) homolog, RAD52 homolog (S. cerevisiae), recombination protein RAD52, rhabdomyosarcoma antigen MU-RMS-40.23 | |
| RAD52 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title