missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD51L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13555-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the RAD51 protein family. RAD51 family members are evolutionarily conserved proteins essential for DNA repair by homologous recombination. This protein has been shown to form a stable heterodimer with the family member RAD51C, which further interacts with the other family members, such as RAD51, XRCC2, and XRCC3. Overexpression of this gene was found to cause cell cycle G1 delay and cell apoptosis, which suggested a role of this protein in sensing DNA damage. At least three alternatively spliced transcript variants encoding distinct isoforms have been observed.Specifications
| RAD51L1 | |
| Polyclonal | |
| Unconjugated | |
| RAD51B | |
| hREC2, R51H2, RAD51 homolog B, RAD51 like 1 (S. cerevisiae), RAD51 like protein 1, RAD51B, RAD51L1, REC2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19363, 500679, 5890 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15315, O35719 | |
| Rad51b | |
| RAD51L1 Fusion Protein Ag4412 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction