Learn More
Invitrogen™ RAD51D Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596713
Description
Immunogen sequence: MGVLRVGLCP GLTEEMIQLL RSHRIKTVVD LVSADLEEVA QKCGLSYKAL VALRRVLLAQ FSAFPVNGAD LYEELKTSTA ILSTGIGSLD KLLDAGLYTG EVTEIVGGPG SGKTQVCLCM AANVAHGLQQ NVLYVDSNGG LTASRLLQLL QAKTQDEEEQ AEALRRIQVV HAFDIFQMLD VLQELRGTVA QQVTGSSGTV KVVVVDSVTA VVSPLLGGQQ REGLALMMQL ARELKTLARD LGMAVVVTNH ITRDRDSGRL KPALGRSWSF VPSTRILLDT IEGAGASGGR RMACLAKSSR QPTGFQEMVD IGTWGTSEQS ATLQGDQT; Positive Samples: Jurkat; Cellular Location: Chromosome, Cytoplasm, Nucleus, centrosome, cytoskeleton, microtubule organizing center, telomere.
Five human RAD 51 homologues have been identified: XRCC2, XRCC3, RAD51B, RAD51C, and RAD51D. Each of these homologues interacts with one or more of the others, with all of the proteins involved in one complex or multiple smaller complexes. RAD51D interacts with XRCC2 and RAD51C. RAD51D's expression pattern and sequence similarity to other RAD51 family members most likely make it part of a complex of proteins required for DNA repair and meiotic recombination.
Specifications
| RAD51D | |
| Polyclonal | |
| Unconjugated | |
| RAD51D | |
| BROVCA4; DNA repair protein RAD51 homolog 4; DNA repair protein RAD51-like 4; HsTRAD; R51H3; RAD51 homolog D; RAD51 paralog D; Rad51d; RAD51L3; RAD51-like 3; RAD51-like protein 3; recombination repair protein; TRAD | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 5892 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.67 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75771 | |
| RAD51D | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-328 of human Rad51D (NP_0028693). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.