missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAD51D Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112682
This item is not returnable.
View return policy
Description
RAD51D Polyclonal Antibody for Western Blot, IHC (P), ELISA
Five human RAD 51 homologues have been identified: XRCC2, XRCC3, RAD51B, RAD51C, and RAD51D. Each of these homologues interacts with one or more of the others, with all of the proteins involved in one complex or multiple smaller complexes. RAD51D interacts with XRCC2 and RAD51C. RAD51D's expression pattern and sequence similarity to other RAD51 family members most likely make it part of a complex of proteins required for DNA repair and meiotic recombination.
Specifications
| RAD51D | |
| Polyclonal | |
| Unconjugated | |
| RAD51D | |
| BROVCA4; DNA repair protein RAD51 homolog 4; DNA repair protein RAD51-like 4; HsTRAD; R51H3; RAD51 homolog D; RAD51 paralog D; Rad51d; RAD51L3; RAD51-like 3; RAD51-like protein 3; recombination repair protein; TRAD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5892 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.82 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75771 | |
| RAD51D | |
| Recombinant Human DNA repair protein RAD51 homolog 4 protein (1-328AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction