missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAD51D Monoclonal Antibody (5B3/6)

Description
Suggested positive control: antigen standard for RAD51L3 (transient overexpression lysate). This antibody does not work on mouse.
Five human RAD 51 homologues have been identified: XRCC2, XRCC3, RAD51B, RAD51C, and RAD51D. Each of these homologues interacts with one or more of the others, with all of the proteins involved in one complex or multiple smaller complexes. RAD51D interacts with XRCC2 and RAD51C. RAD51D's expression pattern and sequence similarity to other RAD51 family members most likely make it part of a complex of proteins required for DNA repair and meiotic recombination.
Specifications
Specifications
| Antigen | RAD51D |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 5B3/6 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.1% sodium azide |
| Gene | RAD51D |
| Gene Accession No. | O55230, O75771 |
| Gene Alias | BROVCA4; DNA repair protein RAD51 homolog 4; DNA repair protein RAD51-like 4; HsTRAD; R51H3; RAD51 homolog D; RAD51 paralog D; Rad51d; RAD51L3; RAD51-like 3; RAD51-like protein 3; recombination repair protein; TRAD |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?