missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD51C Polyclonal Antibody

RAD51C Polyclonal Antibody
Brand: Invitrogen PA575307
This item is not returnable.
View return policy
Description
RAD51C Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| RAD51C | |
| Polyclonal | |
| Unconjugated | |
| RAD51C | |
| BROVCA3; DNA repair protein RAD51 homolog 3; FANCO; MGC104277; R51H3; RAD51 homolog C; RAD51 paralog C; RAD51C; RAD51L2; RAD51-like protein 2; RGD1563765; yeast RAD51 homolog 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114714, 497976, 5889 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43502, Q924H5 | |
| RAD51C | |
| Synthetic peptide corresponding to amino acids 161-210 of Human Rad51C. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction