missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad51C Antibody (Rad51C 2H11/6), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50263IR
This item is not returnable.
View return policy
Description
Rad51C Monoclonal antibody specifically detects Rad51C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| Rad51C | |
| Monoclonal | |
| DyLight 755 | |
| BROVCA3, DNA repair protein RAD51 homolog 3, FANCO, MGC104277, R51H3, RAD51 homolog C, RAD51 homolog C (S. cerevisiae), RAD51L2RAD51 (S. cerevisiae) homolog C, RAD51-like protein 2, yeast RAD51 homolog 3 | |
| Rad51C 2H11/6 was raised against His-tagged human Rad51C, overexpressed in e.coli and purified on a talon affinity column under denaturing conditions, followed by gel purification on a SDS-Page | |
| 0.1 mL | |
| Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination | |
| 5889 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Rad51C 2H11/6 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction