missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad51C Antibody (Rad51C 2H11/6), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50263AF350
This item is not returnable.
View return policy
Description
Rad51C Monoclonal specifically detects Rad51C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Rad51C | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| RAD51C | |
| Rad51C 2H11/6 was raised against His-tagged human Rad51C, overexpressed in e.coli and purified on a talon affinity column under denaturing conditions, followed by gel purification on a SDS-Page | |
| 0.1 mL | |
| 5889 | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Rad51C 2H11/6 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| BROVCA3, DNA repair protein RAD51 homolog 3, FANCO, MGC104277, R51H3, RAD51 homolog C, RAD51 homolog C (S. cerevisiae), RAD51L2RAD51 (S. cerevisiae) homolog C, RAD51-like protein 2, yeast RAD51 homolog 3 | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Rad51C 2H11/6 detects as little as 10ng of purified protein and also detects endogenouse hamster Rad51C in whole V79 cell extracts by western blotting. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction