Learn More
Rad51C Antibody (2H11/6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 29 publications
Brand:  Novus Biologicals NB100-177
 View more versions of this product
Product Code. 18778223
  • £397.00 / 0.10mL
Description
Rad51C Monoclonal specifically detects Rad51C in Human, Mouse, Primate, Yeast samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rad51C
Monoclonal
1 mg/mL
Western Blot 1:1000, Simple Western 1:100, Flow Cytometry 1 ug per million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin reported in scientific literature (PMID 23512992), CyTOF-ready
BROVCA3, DNA repair protein RAD51 homolog 3, FANCO, MGC104277, R51H3, RAD51 homolog C, RAD51 homolog C (S. cerevisiae), RAD51L2RAD51 (S. cerevisiae) homolog C, RAD51-like protein 2, yeast RAD51 homolog 3
Mouse
40 kDa
0.1 mL
Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination
5889
Human, Mouse, Primate, Yeast
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
2H11/6
Unconjugated
O43502
RAD51C
His-tagged human Rad51C, over-expressed in E. coli. [UniProt# O43502]
Protein G purified
RUO
Primary
Does not cross-react with Rad51B, Rad51D, Rad51, XRCC2, or XRCC3 in Western analysis.
Store at 4C. Do not freeze.
IgG1 κ