missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad51C Antibody (2H11/6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 29 publications
Brand: Novus Biologicals NB100-177SS
This item is not returnable.
View return policy
Description
Rad51C Monoclonal specifically detects Rad51C in Human, Mouse, Primate, Yeast samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| Rad51C | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Simple Western 1:100, Flow Cytometry 1 ug per million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin reported in scientific literature (PMID 23512992), CyTOF-ready | |
| BROVCA3, DNA repair protein RAD51 homolog 3, FANCO, MGC104277, R51H3, RAD51 homolog C, RAD51 homolog C (S. cerevisiae), RAD51L2RAD51 (S. cerevisiae) homolog C, RAD51-like protein 2, yeast RAD51 homolog 3 | |
| Mouse | |
| 40 kDa | |
| 0.025 mL | |
| Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination | |
| 5889 | |
| Human, Mouse, Primate, Yeast | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 2H11/6 | |
| Unconjugated | |
| O43502 | |
| RAD51C | |
| His-tagged human Rad51C, over-expressed in E. coli. [UniProt# O43502] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Does not cross-react with Rad51B, Rad51D, Rad51, XRCC2, or XRCC3 in Western analysis. | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction