Learn More
Invitrogen™ RAD51B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA593318
Description
Immunogen sequence: MGSKKLKRVG LSQELCDRLS RHQILTCQDF LCLSPLELMK VTGLSYRGVH ELLCMVSRAC APKMQTAYGI KAQRSADFSP AFLSTTLSAL DEALHGGVAC GSLTEITGPP GCGKTQFCIM MSILATLPTN MGGLEGAVVY IDTESAFSAE RLVEIAESRF PRYFNTEEKL LLTSSKVHLY RELTCDEVLQ RIESLEEEII SKGIKLVILD SVASVVRKEF DAQLQGNLKE RNKFLAREAS SLKYLAEEFS IPVILTNQIT THLSGALASQ ADLVSPADDL SLSEGTSGSS CVIAALGNTW SHSVNTRLIL QYLDSERRQI LIAKSPLAPF TSFVYTIKEE GLVLQETTFC SVTQAELNWA PEILPPQPPE QLGLQMCHHT QLIF; Positive Samples: Mouse kidney, Rat kidney; Cellular Location: Nucleus.
Five human RAD 51 homologues have been identified: XRCC2, XRCC3, RAD51B, RAD51C, and RAD51D. Each of these homologues interacts with one or more of the others, with all of the proteins involved in one complex or multiple smaller complexes. Rad51B plays a role in meiotic recombination and/or recombinational repair.
Specifications
| RAD51B | |
| Polyclonal | |
| Unconjugated | |
| Rad51b | |
| AI553500; DNA repair protein RAD51 homolog 2; hREC2; MGC34245; mREC2; R51H2; R51H2REC2hREC2; RAD51 homolog B; RAD51 paralog B; Rad51B; Rad51l1; RAD51-like 1; RAD51-like protein 1; REC2; RecA-like protein; recombination repair protein; RGD1560681 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19363, 500679 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O35719 | |
| Rad51b | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-384 of human RAD51B (NP_5981932). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.