missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAD51AP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13474887
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13474887 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13474887 Supplier Invitrogen™ Supplier No. PA530395

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, HCT116. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

May participate in a common DNA damage response pathway associated with the activation of homologous recombination and double-strand break repair. Functionally cooperates with PALB2 in promoting of D-loop formation by RAD51. Binds to single and double stranded DNA, and is capable of aggregating DNA. Also binds RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAD51AP1
Applications Western Blot
Classification Polyclonal
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Rad51ap1
Gene Accession No. Q96B01
Gene Alias 2510006L10Rik; PIR51; RAB22; RAD51 associated protein 1; rad51ap1; RAD51-associated protein 1; RAD51-interacting protein
Gene Symbols Rad51ap1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RAD51AP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10635
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.