missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAD51 Recombinant Rabbit Monoclonal Antibody (23GB2275), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | RAD51 |
|---|---|
| Clone | 23GB2275 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089319
![]() |
Invitrogen™
MA553258 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089329
|
Invitrogen™
MA553259 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAD51 Recombinant Monoclonal antibody specifically detects RAD51 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| RAD51 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q06609, Q08297 | |
| 19361, 499870, 5888 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2275 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AV304093; BRCA1/BRCA2-containing complex, subunit 5; BRCC5; DNA repair protein RAD51; DNA repair protein RAD51 homolog 1; DNA repair protein RAD51 homolog A; DNA repair protein rhp51; FANCR; homolog to S.cerevisiae; HRAD51; HsRad51; HsT16930; MGC128559 protein; MRMV2; MUT5; rad51; RAD51 homolog; RAD51 homolog (RecA homolog); RAD51 homolog (RecA homolog, E. coli); RAD51 homolog A; RAD51 recombinase; RAD51 recombinase L homeolog; rad51.1; rad51.L; rad51-1; RAD51A; rad51-a; rad51-b; Reca; RecA family recombinase Rhp51; RecA, E. coli, homolog of; RecA-like protein; recombinase RAD51; recombination protein A; RGD1563603; rhp51; SPAC644.14c; wu:fb38e12; xrad51; XRad51.1; YER095W; zgc:77754 | |
| A synthesized peptide derived from human Rad51 (1-84AA). | |
| Primary | |
| -20°C | |
| Rad51 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title