missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RECA-1 Monoclonal Antibody (HIS52 (87.6E4))
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA123842

Product Code. 11544722

  • £467.00 / 125µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody does not react with mouse, guinea pig, rabbit, porcine, goat, ovine, or chicken protein.

RECA-1 is a cell surface antigen which is expressed by all rat endothelial cells. The endothelium is the thin layer of cells that line the interior surface of blood vessels, forming an interface between circulating blood in the lumen and the rest of the vessel wall. Endothelial cells line the entire circulatory system, from the heart to the smallest capillary.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RECA-1
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide
0
Rad51
Stromal cells from rat lymph node.
125 μg
Primary
Rat
Antibody
IgG1
Immunohistochemistry (Frozen), Immunocytochemistry
HIS52 (87.6E4)
Unconjugated
Rad51
Reca1
Mouse
Protein G
RUO
499870
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.