missing translation for 'onlineSavingsMsg'
Learn More

RAD51 Inhibitor B02, MedChemExpress

Product Code. 16464616
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16464616 100 mg 100mg
16463378 10 mg 10mg
16449906 50 mg 50mg
16426107 5 mg 5mg
16412746 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16464616 Supplier Medchem Express Supplier No. HY101462
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress RAD51 Inhibitor B02 (B02) is an inhibitor of human RAD51 with an IC50 of 27.4 μM.

RAD51 Inhibitor B02 (B02) is an inhibitor of human RAD51 with an IC50 of 27.4 μM.

Specifications

Chemical Name or Material RAD51 Inhibitor B02
Physical Form Powder
CAS 1290541-46-6
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C22H17N3O
For Use With (Application) Cancer-programmed cell death
Synonym B02
Solubility Information DMSO : ≥ 37 mg/mL (109.02 mM)
SMILES O=C1N(CC2=CC=CC=C2)C(/C=C/C3=CC=CN=C3)=NC4=C1C=CC=C4
Molecular Weight (g/mol) 339.39
Formula Weight 339.39
Percent Purity 99.69%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.