missing translation for 'onlineSavingsMsg'
Learn More

Cabotegravir, MedChemExpress

Product Code. 16412747
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16412747

Brand: Medchem Express HY15592

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cabotegravir (GSK-1265744) is a orally active and long-acting HIV integrase inhibitor and organic anion transporter 1/3 (OAT1/OAT3) inhibitor with IC50 values of 2.5 nM, 0.41 μM and 0.81 μM for HIVADA, OAT3 and OAT1, respectively.

Cabotegravir is primarily metabolized by uridine diphosphate glucuronosyltransferase (UGT) 1A1, with low potential to interact with other antiretroviral drugs (ARVs). Cabotegravir can be used to research AIDS.

Specifications

Chemical Name or Material Cabotegravir
Physical Form Powder
CAS 1051375-10-0
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H17F2N3O5
For Use With (Application) COVID-19-anti-virus
Synonym GSK-1265744 S/GSK1265744
Solubility Information DMSO : 16.67 mg/mL (41.12 mM; Need ultrasonic)
SMILES FC1=C(CNC(C2=CN3C(C(N([C@@H](C)CO4)[C@@]4([H])C3)=O)=C(O)C2=O)=O)C=CC(F)=C1
Molecular Weight (g/mol) 405.35
Formula Weight 405.35
Percent Purity 98.04%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.