missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad51 Antibody (RAD51/2701), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79895MFV500
This item is not returnable.
View return policy
Description
Rad51 Monoclonal antibody specifically detects Rad51 in Human samples. It is validated for Flow Cytometry
Specifications
| Rad51 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| RAD51/2701 | |
| Flow Cytometry | |
| BRCC5, HRAD51, HsRAD51, HsT16930, RAD51 (S. cerevisiae) homolog (E coli RecA homolog), RAD51 homolog (RecA homolog, E. coli) (S. cerevisiae), RAD51 homolog A, RAD51ABRCA1/BRCA2-containing complex, subunit 5, RAD51L3, RecA, E. coli, homolog of, RECADNA repair protein RAD51 homolog 1, RecA-like protein, recombination protein A | |
| Recombinant fragment of human Rad51 protein (around aa 1-134) (exact sequence is proprietary) (Uniprot: Q06609) | |
| 0.1 mL | |
| Cancer, DNA Repair, Homologous Recombination, modENCODE Antibodies, Phospho Specific | |
| 5888 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction