missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad51 Antibody (523713), Unconjugated, Novus Biologicals™
Mouse Monoclonal Antibody
£193.00 - £417.00
Specifications
| Antigen | Rad51 |
|---|---|
| Clone | 523713 |
| Concentration | 1.0 mg/mL |
| Dilution | Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 5 - 10 μg/mL |
| Applications | Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18615544
|
Novus Biologicals
MAB99481 |
100 μg |
£417.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18652504
|
Novus Biologicals
MAB99481-SP |
25 μg |
£193.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rad51 Monoclonal antibody specifically detects Rad51 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Rad51 | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, DNA Repair, Homologous Recombination, modENCODE Antibodies | |
| PBS | |
| 5888 | |
| IgG3 | |
| Protein A or G purified |
| 523713 | |
| Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 5 - 10 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BRCC5, HRAD51, HsRAD51, HsT16930, RAD51 (S. cerevisiae) homolog (E coli RecA homolog), RAD51 homolog (RecA homolog, E. coli) (S. cerevisiae), RAD51 homolog A, RAD51ABRCA1/BRCA2-containing complex, subunit 5, RAD51L3, RecA, E. coli, homolog of, RECADNA repair protein RAD51 homolog 1, RecA-like protein, recombination protein A | |
| E. coli-derived partial recombinant human Rad51 protein (amino acids 174-339) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title