missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad51 Antibody (523713), DyLight 488, R&D Systems™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB99481G
This item is not returnable.
View return policy
Description
Rad51 Monoclonal specifically detects Rad51 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Rad51 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RAD51 | |
| E. coli-derived partial recombinant human Rad51 protein (amino acids 174-339) | |
| 0.1 mL | |
| Cancer, DNA Repair, Homologous Recombination, modENCODE Antibodies, Phospho Specific | |
| Human | |
| IgG3 |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 523713 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| BRCC5, HRAD51, HsRAD51, HsT16930, RAD51 (S. cerevisiae) homolog (E coli RecA homolog), RAD51 homolog (RecA homolog, E. coli) (S. cerevisiae), RAD51 homolog A, RAD51ABRCA1/BRCA2-containing complex, subunit 5, RAD51L3, RecA, E. coli, homolog of, RECADNA repair protein RAD51 homolog 1, RecA-like protein, recombination protein A | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction