missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad50 Antibody (13B3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 78 publications
Brand: Novus Biologicals NB100-147-0.1ml
This item is not returnable.
View return policy
Description
Rad50 Monoclonal specifically detects Rad50 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, In vitro assay, Knockdown Validated.
Specifications
| Rad50 | |
| Monoclonal | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| RAD50 | |
| Amino acids 1-425 of Rad50 expressed in E. coli. | |
| Protein G purified | |
| RUO | |
| Primary | |
| This is specific for Rad50. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 13B3 | |
| Unconjugated | |
| DNA repair protein RAD50, EC 3.6, EC 3.6.1.15, EC 3.6.3, EC 3.6.3.27, hRAD50, NBSLD, RAD50 (S. cerevisiae) homolog, RAD50 homolog (S. cerevisiae), RAD502, RAD50-2 | |
| Mouse | |
| 153 kDa | |
| 0.1 mL | |
| Breast Cancer, DNA Repair, Homologous Recombination | |
| 10111 | |
| Human, Mouse, Rat, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction