missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAD3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA532174

Product Code. 13466547

  • £440.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: Yeast lysate. Predicted reactivity: Yeast (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ATP-dependent DNA helicase involved in excision repair of DNA damaged with UV light, bulky adducts, or cross-linking agents. Necessary for excision of pyrimidine dimers. Also unwinds DNA/RNA duplexes. Plays an essential role in the cell viability. Involved in the maintenance of the fidelity of DNA replication. Acts as component of the general transcription and DNA repair factor IIH (TFIIH) core, which is essential for both basal and activated transcription, and is involved in nucleotide excision repair (NER) of damaged DNA. TFIIH has CTD kinase and DNA-dependent ATPase activity, and is essential for polymerase II transcription in vitro.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RAD3
Polyclonal
Unconjugated
RAD3
DNA repair helicase RAD3; General transcription and DNA repair factor IIH helicase subunit XPD; RAD3; REM1; RNA polymerase II transcription factor B subunit RAD3; TFB subunit RAD3; TFIIH subunit XPD; TFIIH/NER complex ATP-dependent 5'-3' DNA helicase subunit RAD3; YER171W
Rabbit
Antigen affinity chromatography
RUO
856918
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.9 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
P06839
RAD3
Recombinant protein encompassing a sequence within the N-terminus region of Saccharomyces cerevisiae S288c RAD3. The exact sequence is proprietary.
100 μL
Primary
Yeast
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.